The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308388.1 | NP_001295317.1 | 12 - 13 | 965 | 88 |
NM_153635.2 | NP_705899.2 | 12 - 13 | 946 | 88 | |
XM_011533386.2 | XP_011531688.1 | 11 - 12 | 1122 | 88 | |
XM_011533388.2 | XP_011531690.1 | 11 - 12 | 1122 | 88 | |
XM_011533389.2 | XP_011531691.1 | 11 - 12 | 1122 | 88 | |
GenBank mRNA | BC035735.1 | - | 12 - 13 | 946 | 88 |
BC130375.1 | - | 12 - 13 | 817 | 88 | |
BX453546.2 | - | 4 - 5 | 88 |