The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142643.2 | NP_001136115.1 | 6 - 7 | 684 | 77 |
NM_020753.4 | NP_065804.2 | 7 - 8 | 1201 | 77 | |
GenBank mRNA | AB032965.1 | - | 3 - 4 | 380 | 77 |
AF451976.1 | - | 7 - 8 | 1054 | 77 | |
AK300354.1 | - | 6 - 7 | 631 | 77 | |
AK315936.1 | - | 6 - 7 | 685 | 77 | |
BC066643.1 | - | 7 - 8 | 1164 | 77 |