The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031958.1 | NP_114164.1 | 1 - 1 | 56 | 93 |
GenBank mRNA | AJ406931.1 | - | 1 - 1 | 56 | 93 |
BC113077.1 | - | 1 - 1 | 36 | 93 | |
BC113078.1 | - | 1 - 1 | 40 | 93 |