The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145307.4 | NP_001138779.1 | 3 - 4 | 773 | 67 |
NM_001145309.3 | NP_001138781.1 | 3 - 4 | 773 | 67 | |
NM_001145310.3 | NP_001138782.1 | 3 - 4 | 773 | 67 | |
NM_001271471.2 | NP_001258400.1 | 3 - 4 | 773 | 67 | |
NM_001318803.1 | NP_001305732.1 | 3 - 4 | 730 | 67 | |
NM_145309.5 | NP_660352.1 | 3 - 4 | 773 | 67 | |
XM_006718473.3 | XP_006718536.1 | 3 - 4 | 999 | 67 | |
XM_006718474.3 | XP_006718537.1 | 2 - 3 | 1038 | 67 | |
XM_011544847.2 | XP_011543149.1 | 3 - 4 | 1227 | 67 | |
XM_011544848.2 | XP_011543150.1 | 4 - 5 | 1285 | 67 | |
XM_017017359.1 | XP_016872848.1 | 4 - 5 | 1390 | 67 | |
XM_017017360.1 | XP_016872849.1 | 4 - 5 | 1242 | 67 | |
GenBank mRNA | AK313150.1 | - | 3 - 4 | 457 | 67 |
BC012855.1 | - | 3 - 4 | 245 | 67 | |
BI461207.1 | - | 3 - 4 | 216 | 67 | |
EU627066.1 | - | 3 - 4 | 245 | 67 | |
EU627067.1 | - | 2 - 3 | 195 | 67 | |
EU627068.1 | - | 3 - 4 | 245 | 67 | |
EU627069.1 | - | 3 - 4 | 245 | 67 | |
EU627070.1 | - | 3 - 4 | 245 | 67 |