Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD18A
Entrez Gene ID
253650
Gene Name
ankyrin repeat domain 18A
Gene Aliases
-
Chromosome Location
Chr.9: 38566260 - 38620587 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006716749.3 XP_006716812.1 16 - 17 3320 152
XM_011517836.2 XP_011516138.1 16 - 17 3321 152
XM_011517837.2 XP_011516139.1 16 - 17 3321 152
GenBank mRNA AK093059.1 - 2 - 3 379 152
AY550933.1 - 2 - 3 328 152
BC131497.1 - 2 - 3 375 152

View Full Product Details