The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177428.1 | NP_001170899.1 | 10 - 11 | 1784 | 89 |
NM_001177431.1 | NP_001170902.1 | 12 - 13 | 1792 | 89 | |
NM_001177433.1 | NP_001170904.1 | 9 - 10 | 1604 | 89 | |
NM_021625.4 | NP_067638.3 | 12 - 13 | 1983 | 89 | |
NM_147204.2 | NP_671737.1 | 10 - 11 | 1745 | 89 | |
XM_005253918.1 | XP_005253975.1 | 12 - 13 | 2343 | 89 | |
XM_011538630.2 | XP_011536932.2 | 12 - 13 | 3086 | 89 | |
XM_011538631.2 | XP_011536933.2 | 11 - 12 | 2946 | 89 | |
XM_011538632.2 | XP_011536934.2 | 11 - 12 | 2907 | 89 | |
XM_011538633.2 | XP_011536935.2 | 10 - 11 | 2766 | 89 | |
XM_011538634.2 | XP_011536936.2 | 12 - 13 | 3088 | 89 | |
XM_017019774.1 | XP_016875263.1 | 12 - 13 | 2716 | 89 | |
GenBank mRNA | AB032427.1 | - | 11 - 12 | 1989 | 89 |
AB073669.1 | - | 10 - 11 | 1714 | 89 | |
AB100308.1 | - | 11 - 12 | 1894 | 89 | |
AF258465.1 | - | 11 - 12 | 1894 | 89 | |
AF279673.1 | - | 11 - 12 | 1690 | 89 | |
AJ296305.1 | - | 12 - 13 | 1983 | 89 | |
AL834192.1 | - | 12 - 13 | 1954 | 90 | |
BC117426.1 | - | 12 - 13 | 1977 | 89 | |
BC143307.1 | - | 12 - 13 | 2019 | 89 | |
BC143315.1 | - | 12 - 13 | 1977 | 89 | |
DQ059644.1 | - | 10 - 11 | 1753 | 89 | |
DQ059645.1 | - | 12 - 13 | 1792 | 89 | |
DQ059646.1 | - | 9 - 10 | 1573 | 89 |