The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152701.4 | NP_689914.3 | 53 - 54 | 14077 | 64 |
XM_011515130.2 | XP_011513432.1 | 53 - 54 | 14112 | 64 | |
XM_011515131.2 | XP_011513433.1 | 53 - 54 | 14019 | 64 | |
XM_011515132.2 | XP_011513434.1 | 52 - 53 | 14019 | 64 | |
XM_011515133.2 | XP_011513435.1 | 52 - 53 | 13974 | 64 | |
XM_011515134.2 | XP_011513436.1 | 53 - 54 | 14112 | 64 | |
XM_011515136.2 | XP_011513438.2 | 48 - 49 | 13477 | 64 | |
GenBank mRNA | AF501281.1 | - | 35 - 36 | 5260 | 64 |
AK091270.1 | - | 1 - 2 | 80 | 64 | |
AK128516.1 | - | 10 - 11 | 1372 | 64 | |
AK131360.1 | - | 19 - 20 | 2849 | 64 | |
AY204751.1 | - | 53 - 54 | 14078 | 64 | |
JF913488.1 | - | 36 - 37 | 5975 | 64 | |
JF913491.1 | - | 36 - 37 | 5882 | 64 | |
JF913492.1 | - | 36 - 37 | 5975 | 64 |