The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040272.5 | NP_001035362.3 | 24 - 25 | 4612 | 98 |
XM_011518063.2 | XP_011516365.1 | 26 - 27 | 4750 | 98 | |
XM_011518064.2 | XP_011516366.1 | 25 - 26 | 4865 | 98 | |
XM_011518067.1 | XP_011516369.1 | 16 - 17 | 3480 | 98 | |
XM_011518068.2 | XP_011516370.1 | 14 - 15 | 4083 | 98 | |
XM_011518070.2 | XP_011516372.1 | 14 - 15 | 3248 | 98 | |
XM_017015310.1 | XP_016870799.1 | 25 - 26 | 11387 | 98 | |
XM_017015311.1 | XP_016870800.1 | 25 - 26 | 4699 | 98 | |
XM_017015312.1 | XP_016870801.1 | 24 - 25 | 4815 | 98 | |
XM_017015313.1 | XP_016870802.1 | 25 - 26 | 4513 | 98 | |
GenBank mRNA | AK092602.1 | - | 3 - 4 | 671 | 98 |
AK123028.1 | - | 1 - 2 | 97 | 98 | |
BC100790.2 | - | 3 - 4 | 660 | 98 | |
BX640951.1 | - | 5 - 6 | 745 | 98 |