Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX1
Entrez Gene ID
6642
Gene Name
sorting nexin 1
Gene Aliases
HsT17379, VPS5
Chromosome Location
Chr.15: 64094071 - 64144234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242933.1 NP_001229862.1 9 - 10 1054 80
NM_003099.4 NP_003090.2 9 - 10 1054 80
NM_148955.3 NP_683758.1 7 - 8 859 80
XM_005254611.2 XP_005254668.1 9 - 10 1030 80
GenBank mRNA AB209013.1 - 9 - 10 861 80
AF065483.1 - 9 - 10 924 80
AF065484.1 - 7 - 8 729 80
AK128179.1 - 9 - 10 946 80
AK222561.1 - 9 - 10 938 80
AK222793.1 - 9 - 10 927 80
AK291752.1 - 9 - 10 938 80
AK310618.1 - 6 - 7 738 80
BC000357.2 - 9 - 10 934 80
BT006983.1 - 9 - 10 924 80
U53225.1 - 9 - 10 969 80

View Full Product Details