The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305121.1 | NP_001292050.1 | 4 - 5 | 1109 | 84 |
NM_001305122.1 | NP_001292051.1 | 5 - 6 | 1174 | 84 | |
NM_153208.2 | NP_694940.1 | 5 - 6 | 1174 | 84 | |
XM_011545736.2 | XP_011544038.1 | 4 - 5 | 509 | 84 | |
XM_017022933.1 | XP_016878422.1 | 4 - 5 | 1220 | 84 | |
GenBank mRNA | AF520570.1 | - | 5 - 6 | 1174 | 84 |
AK000122.1 | - | 4 - 5 | 1109 | 84 | |
AK301933.1 | - | 4 - 5 | 497 | 84 | |
AK303998.1 | - | 3 - 4 | 402 | 84 | |
AK315672.1 | - | 4 - 5 | 497 | 84 | |
BC034823.1 | - | 4 - 5 | 505 | 84 |