The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039876.2 | NP_001034965.1 | 1 - 2 | 244 | 101 |
NM_001297735.2 | NP_001284664.1 | 1 - 2 | 244 | 101 | |
GenBank mRNA | AK093764.1 | - | 1 - 2 | 218 | 101 |
BC052573.1 | - | 1 - 2 | 234 | 101 |