The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039876.2 | NP_001034965.1 | 4 - 5 | 733 | 100 |
GenBank mRNA | AK093764.1 | - | 4 - 5 | 707 | 100 |