The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153270.2 | NP_695002.1 | 1 - 1 | 207 | 94 |
GenBank mRNA | AK092279.1 | - | 1 - 1 | 150 | 94 |
BC109031.2 | - | 1 - 1 | 119 | 94 | |
DA395149.1 | - | 1 - 1 | 207 | 94 |