Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFPM1
Entrez Gene ID
161882
Gene Name
zinc finger protein, FOG family member 1
Gene Aliases
FOG, FOG1, ZC2HC11A, ZNF408, ZNF89A
Chromosome Location
Chr.16: 88451782 - 88535166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153813.2 NP_722520.2 2 - 3 179 122
XM_011522912.2 XP_011521214.1 2 - 3 4028 122
XM_011522914.2 XP_011521216.1 2 - 3 263 122
GenBank mRNA AF488691.1 - 2 - 3 468 122
BE219988.1 - 2 - 3 179 122

View Full Product Details