Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAPBP
Entrez Gene ID
6892
Gene Name
TAP binding protein (tapasin)
Gene Aliases
NGS17, TAPA, TPN, TPSN
Chromosome Location
Chr.6: 33299694 - 33314387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003190.4 NP_003181.3 3 - 4 811 96
NM_172208.2 NP_757345.2 3 - 4 811 96
XM_011514828.2 XP_011513130.1 3 - 4 678 96
XM_017011227.1 XP_016866716.1 3 - 4 678 96
GenBank mRNA AB010639.1 - 3 - 4 478 96
AF009510.1 - 3 - 4 481 96
AF029750.1 - 3 - 4 503 96
AF067286.3 - 3 - 4 473 96
AF314222.1 - 3 - 4 465 96
BC080574.1 - 3 - 4 636 96

View Full Product Details