The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282489.2 | NP_001269418.1 | 2 - 3 | 1521 | 93 |
NM_173477.4 | NP_775748.2 | 2 - 3 | 1574 | 93 | |
XM_011524296.2 | XP_011522598.1 | 2 - 3 | 1547 | 93 | |
GenBank mRNA | AK091243.1 | - | 2 - 3 | 1567 | 93 |
AK289804.1 | - | 2 - 3 | 1571 | 93 | |
AK296899.1 | - | 2 - 3 | 1514 | 93 | |
BC101096.2 | - | 2 - 3 | 1461 | 93 | |
BC101097.1 | - | 2 - 3 | 1461 | 93 | |
BC101098.2 | - | 2 - 3 | 1461 | 93 | |
BC101099.1 | - | 2 - 3 | 1461 | 93 |