The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136160.1 | NP_001129632.1 | 9 - 10 | 1211 | 142 |
NM_001136161.1 | NP_001129633.1 | 9 - 10 | 1246 | 142 | |
NM_173518.4 | NP_775789.3 | 9 - 10 | 1246 | 142 | |
XM_005251174.2 | XP_005251231.1 | 8 - 9 | 1108 | 142 | |
XM_006716427.3 | XP_006716490.1 | 9 - 10 | 1184 | 142 | |
XM_006716433.3 | XP_006716496.1 | 7 - 8 | 926 | 142 | |
XM_011517467.2 | XP_011515769.1 | 9 - 10 | 1205 | 142 | |
XM_011517468.2 | XP_011515770.1 | 8 - 9 | 1079 | 142 | |
XM_011517469.2 | XP_011515771.1 | 9 - 10 | 1205 | 142 | |
GenBank mRNA | AK098558.1 | - | 5 - 6 | 782 | 142 |
AK302169.1 | - | 9 - 10 | 1211 | 142 | |
AK302672.1 | - | 9 - 10 | 1205 | 142 | |
AK303003.1 | - | 7 - 8 | 1002 | 142 | |
AK304170.1 | - | 8 - 9 | 1108 | 142 | |
BC034576.1 | - | 9 - 10 | 1246 | 142 | |
BC063108.1 | - | 8 - 9 | 1124 | 142 |