Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNC5CL
Entrez Gene ID
222643
Gene Name
unc-5 family C-terminal like
Gene Aliases
MUXA, ZUD
Chromosome Location
Chr.6: 41026901 - 41039199 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173561.2 NP_775832.2 7 - 8 1320 70
GenBank mRNA AY510109.1 - 6 - 7 1221 70
BC035284.1 - 7 - 8 1310 70
BM312066.1 - 4 - 5 362 70
JF681947.1 - 7 - 8 1338 70

View Full Product Details