The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173561.2 | NP_775832.2 | 8 - 9 | 1431 | 127 |
GenBank mRNA | AY510109.1 | - | 7 - 8 | 1332 | 127 |
BC035284.1 | - | 8 - 9 | 1421 | 127 | |
JF681947.1 | - | 8 - 9 | 1449 | 127 |