The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173598.4 | NP_775869.3 | 12 - 13 | 1764 | 86 |
XM_011538224.2 | XP_011536526.1 | 12 - 13 | 2093 | 86 | |
XM_011538225.2 | XP_011536527.1 | 12 - 13 | 2558 | 86 | |
XM_011538226.2 | XP_011536528.1 | 12 - 13 | 2099 | 86 | |
XM_011538228.2 | XP_011536530.1 | 10 - 11 | 923 | 86 | |
XM_011538229.2 | XP_011536531.1 | 12 - 13 | 2099 | 86 | |
XM_017019208.1 | XP_016874697.1 | 12 - 13 | 2099 | 86 | |
XM_017019210.1 | XP_016874699.1 | 7 - 8 | 549 | 86 | |
GenBank mRNA | AK098831.1 | - | 10 - 11 | 942 | 86 |
AY345972.1 | - | 10 - 11 | 1433 | 86 | |
BC107106.1 | - | 10 - 11 | 1428 | 86 | |
BC107107.1 | - | 10 - 11 | 1428 | 86 | |
BC127603.1 | - | 10 - 11 | 1433 | 86 |