Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA3A
Entrez Gene ID
146177
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
-
Chromosome Location
Chr.16: 22091930 - 22156966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK098260.1 - 17 - 18 1931 79

View Full Product Details