The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321399.1 | NP_001308328.1 | 3 - 4 | 501 | 84 |
GenBank mRNA | AK096966.1 | - | 3 - 4 | 502 | 84 |