Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCR2
Entrez Gene ID
9436
Gene Name
natural cytotoxicity triggering receptor 2
Gene Aliases
CD336, LY95, NK-p44, NKP44, dJ149M18.1
Chromosome Location
Chr.6: 41335620 - 41350887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199510.1 NP_001186439.1 3 - 4 618 92
NM_004828.3 NP_004819.2 3 - 4 618 92
XM_017011500.1 XP_016866989.1 3 - 4 788 92
GenBank mRNA AJ010099.1 - 3 - 4 557 92

View Full Product Details