The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025003.3 | NP_079279.3 | 1 - 2 | 93 | 111 |
XM_011538754.2 | XP_011537056.1 | 1 - 2 | 143 | 111 | |
GenBank mRNA | AF488804.1 | - | 1 - 2 | 93 | 111 |
AJ515153.3 | - | 1 - 2 | 93 | 111 | |
AJ515154.2 | - | 1 - 2 | 93 | 111 |