Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS20
Entrez Gene ID
80070
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 20
Gene Aliases
ADAM-TS20, ADAMTS-20, GON-1
Chromosome Location
Chr.12: 43352843 - 43551971 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025003.3 NP_079279.3 25 - 26 3654 100
XM_011538754.2 XP_011537056.1 25 - 26 3707 100
XM_017019979.1 XP_016875468.1 18 - 19 2723 100
GenBank mRNA AF488804.1 - 25 - 26 3657 100
AJ515153.3 - 25 - 26 3648 100
AJ515154.2 - 25 - 26 3648 100

View Full Product Details