The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291728.1 | NP_001278657.1 | 2 - 3 | 331 | 70 |
NM_001291730.1 | NP_001278659.1 | 2 - 3 | 331 | 70 | |
NM_001291732.1 | NP_001278661.1 | 2 - 3 | 331 | 70 | |
NM_001291733.1 | NP_001278662.1 | 2 - 3 | 331 | 70 | |
NM_033419.4 | NP_219487.3 | 2 - 3 | 331 | 70 | |
XM_011525480.2 | XP_011523782.1 | 2 - 3 | 318 | 70 | |
XM_011525481.2 | XP_011523783.1 | 2 - 3 | 318 | 70 | |
GenBank mRNA | AB088396.1 | - | 2 - 3 | 286 | 70 |
AK057911.1 | - | 3 - 4 | 297 | 70 | |
AK294639.1 | - | 2 - 3 | 318 | 70 | |
AK301106.1 | - | 3 - 4 | 233 | 70 | |
AK314469.1 | - | 2 - 3 | 291 | 70 | |
AY358437.1 | - | 2 - 3 | 307 | 70 | |
BC010652.1 | - | 2 - 3 | 292 | 70 | |
DC389056.1 | - | 2 - 3 | 332 | 70 |