Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAPD4
Entrez Gene ID
167153
Gene Name
PAP associated domain containing 4
Gene Aliases
GLD2, TUT2
Chromosome Location
Chr.5: 79612416 - 79686648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114393.1 NP_001107865.1 3 - 4 562 109
NM_001114394.1 NP_001107866.1 2 - 3 837 109
NM_001297744.1 NP_001284673.1 3 - 4 562 109
NM_001297745.1 NP_001284674.1 2 - 3 837 109
NM_173797.3 NP_776158.2 3 - 4 596 109
XM_011543219.2 XP_011541521.1 2 - 3 841 109
XM_011543220.2 XP_011541522.1 3 - 4 911 109
XM_011543221.2 XP_011541523.1 4 - 5 1000 109
XM_011543222.2 XP_011541524.1 3 - 4 930 109
XM_011543223.2 XP_011541525.1 3 - 4 550 109
XM_011543224.2 XP_011541526.1 3 - 4 549 109
XM_011543225.2 XP_011541527.1 4 - 5 511 109
XM_011543226.2 XP_011541528.1 4 - 5 631 109
XM_011543227.2 XP_011541529.1 5 - 6 720 109
XM_011543228.2 XP_011541530.1 2 - 3 841 109
XM_011543229.2 XP_011541531.1 2 - 3 841 109
XM_011543230.2 XP_011541532.1 2 - 3 841 109
XM_011543231.2 XP_011541533.1 2 - 3 841 109
XM_017009151.1 XP_016864640.1 4 - 5 988 109
XM_017009152.1 XP_016864641.1 3 - 4 911 109
XM_017009153.1 XP_016864642.1 3 - 4 911 109
XM_017009154.1 XP_016864643.1 4 - 5 988 109
XM_017009155.1 XP_016864644.1 3 - 4 930 109
XM_017009156.1 XP_016864645.1 4 - 5 1220 109
XM_017009157.1 XP_016864646.1 2 - 3 841 109
XM_017009158.1 XP_016864647.1 3 - 4 911 109
XM_017009159.1 XP_016864648.1 3 - 4 930 109
XM_017009160.1 XP_016864649.1 4 - 5 511 109
XM_017009161.1 XP_016864650.1 3 - 4 930 109
XM_017009162.1 XP_016864651.1 3 - 4 518 109
GenBank mRNA AK095818.1 - 3 - 4 443 109
AL833136.1 - 2 - 3 823 109
BC026061.1 - 4 - 5 595 109
BC047581.1 - 3 - 4 541 109
CN348312.1 - 3 - 4 451 109

View Full Product Details