The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190201.1 | NP_001177130.1 | 11 - 12 | 1382 | 59 |
NM_001318506.1 | NP_001305435.1 | 11 - 12 | 1438 | 59 | |
XM_005255896.4 | XP_005255953.1 | 13 - 14 | 1698 | 59 | |
XM_011523021.2 | XP_011521323.1 | 14 - 15 | 1830 | 59 | |
XM_011523023.2 | XP_011521325.1 | 14 - 15 | 1818 | 59 | |
XM_011523025.2 | XP_011521327.1 | 13 - 14 | 1749 | 59 | |
XM_011523026.2 | XP_011521328.1 | 13 - 14 | 1710 | 59 | |
XM_011523027.2 | XP_011521329.1 | 13 - 14 | 1701 | 59 | |
XM_011523028.2 | XP_011521330.1 | 13 - 14 | 1718 | 59 | |
XM_011523030.2 | XP_011521332.1 | 11 - 12 | 1488 | 59 | |
XM_017023160.1 | XP_016878649.1 | 13 - 14 | 1737 | 59 | |
XM_017023161.1 | XP_016878650.1 | 13 - 14 | 1689 | 59 | |
XM_017023163.1 | XP_016878652.1 | 12 - 13 | 1620 | 59 | |
XM_017023164.1 | XP_016878653.1 | 12 - 13 | 1608 | 59 | |
GenBank mRNA | AK094783.1 | - | 11 - 12 | 1399 | 59 |
AK293061.1 | - | 11 - 12 | 1230 | 59 | |
AK300792.1 | - | 11 - 12 | 1342 | 59 | |
BC064573.1 | - | 11 - 12 | 1162 | 59 | |
BC084555.1 | - | 10 - 11 | 1196 | 59 |