Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
Chromosome Location
Chr.14: 99511266 - 99604390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144995.1 NP_001138467.1 6 - 6 2126 62
XM_011536705.2 XP_011535007.1 7 - 7 1809 62
GenBank mRNA AK093969.1 - 1 - 1 269 62
AK123840.1 - 5 - 5 1014 62
BC110042.1 - 1 - 1 392 62

View Full Product Details