Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBX7
Entrez Gene ID
23492
Gene Name
chromobox 7
Gene Aliases
-
Chromosome Location
Chr.22: 39130772 - 39152661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175709.3 NP_783640.1 4 - 5 336 54
XM_006724174.3 XP_006724237.1 4 - 5 482 54
XM_006724175.3 XP_006724238.1 4 - 5 484 54
XM_017028679.1 XP_016884168.1 4 - 5 458 54

View Full Product Details