Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3582423-3582696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5629075 c.5355G>C p.L1785L Chr.16: 3582492-3582492 on Build GRCh38
127452 c.5238C>T p.A1746A Chr.16: 3582609-3582609 on Build GRCh38
4728186 c.5246C>T p.A1749V Chr.16: 3582601-3582601 on Build GRCh38
4818714 c.5165G>C p.C1722S Chr.16: 3582682-3582682 on Build GRCh38
4060645 c.5192C>T p.A1731V Chr.16: 3582655-3582655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746396836 Chr.16: 3582502-3582502 on Build GRCh38
rs138445982 Chr.16: 3582579-3582579 on Build GRCh38
rs770529698 Chr.16: 3582461-3582461 on Build GRCh38
rs539125446 Chr.16: 3582653-3582653 on Build GRCh38
rs562491450 Chr.16: 3582493-3582493 on Build GRCh38
rs767246431 Chr.16: 3582473-3582473 on Build GRCh38
rs762619200 Chr.16: 3582464-3582464 on Build GRCh38
rs752111888 Chr.16: 3582625-3582625 on Build GRCh38
rs551495566 Chr.16: 3582562-3582562 on Build GRCh38
rs577770118 Chr.16: 3582444-3582444 on Build GRCh38
rs1011190951 Chr.16: 3582609-3582609 on Build GRCh38
rs770569508 Chr.16: 3582539-3582539 on Build GRCh38
rs369714623 Chr.16: 3582633-3582633 on Build GRCh38
rs528519471 Chr.16: 3582610-3582610 on Build GRCh38
rs764337343 Chr.16: 3582480-3582480 on Build GRCh38
rs973283650 Chr.16: 3582512-3582512 on Build GRCh38
rs904241737 Chr.16: 3582548-3582548 on Build GRCh38
rs372921011 Chr.16: 3582583-3582583 on Build GRCh38
rs200262630 Chr.16: 3582624-3582624 on Build GRCh38
rs569225834 Chr.16: 3582639-3582639 on Build GRCh38
rs147251446 Chr.16: 3582667-3582667 on Build GRCh38
rs1031274414 Chr.16: 3582556-3582556 on Build GRCh38
rs374876624 Chr.16: 3582482-3582482 on Build GRCh38
rs144057906 Chr.16: 3582619-3582619 on Build GRCh38
rs907816292 Chr.16: 3582497-3582497 on Build GRCh38
rs747442815 Chr.16: 3582520-3582520 on Build GRCh38
rs984137258 Chr.16: 3582620-3582620 on Build GRCh38
rs780261532 Chr.16: 3582511-3582511 on Build GRCh38
rs769567874 Chr.16: 3582529-3582529 on Build GRCh38
rs144776083 Chr.16: 3582588-3582588 on Build GRCh38
rs201214017 Chr.16: 3582608-3582608 on Build GRCh38
rs560381786 Chr.16: 3582475-3582475 on Build GRCh38
rs754067610 Chr.16: 3582575-3582575 on Build GRCh38
rs796844929 Chr.16: 3582541-3582541 on Build GRCh38
rs971291316 Chr.16: 3582446-3582446 on Build GRCh38
rs61031231 Chr.16: 3582630-3582630 on Build GRCh38
rs759412447 Chr.16: 3582558-3582558 on Build GRCh38
rs377329498 Chr.16: 3582601-3582601 on Build GRCh38
rs759307727 Chr.16: 3582470-3582470 on Build GRCh38
rs542981864 Chr.16: 3582483-3582483 on Build GRCh38
rs376960842 Chr.16: 3582658-3582658 on Build GRCh38
rs371875095 Chr.16: 3582476-3582476 on Build GRCh38
rs369180077 Chr.16: 3582554-3582554 on Build GRCh38
rs143818824 Chr.16: 3582566-3582566 on Build GRCh38
rs981311587 Chr.16: 3582590-3582590 on Build GRCh38
rs769157164 Chr.16: 3582458-3582459 on Build GRCh38
rs753159332 Chr.16: 3582638-3582638 on Build GRCh38
rs752633436 Chr.16: 3582670-3582670 on Build GRCh38
rs781479923 Chr.16: 3582617-3582618 on Build GRCh38
rs1049534327 Chr.16: 3582545-3582545 on Build GRCh38
rs775552034 Chr.16: 3582600-3582600 on Build GRCh38
rs1041339962 Chr.16: 3582492-3582492 on Build GRCh38
rs140231739 Chr.16: 3582618-3582618 on Build GRCh38
rs748749970 Chr.16: 3582532-3582532 on Build GRCh38
rs199838670 Chr.16: 3582659-3582659 on Build GRCh38
rs140666540 Chr.16: 3582669-3582669 on Build GRCh38
rs755406950 Chr.16: 3582587-3582587 on Build GRCh38
rs750467283 Chr.16: 3582614-3582614 on Build GRCh38
rs780678810 Chr.16: 3582584-3582584 on Build GRCh38
rs370495748 Chr.16: 3582597-3582597 on Build GRCh38
rs776789371 Chr.16: 3582605-3582605 on Build GRCh38
rs761845224 Chr.16: 3582576-3582576 on Build GRCh38
rs748501466 Chr.16: 3582624-3582627 on Build GRCh38
rs1029761357 Chr.16: 3582641-3582641 on Build GRCh38
rs774442734 Chr.16: 3582466-3582466 on Build GRCh38
rs878855163 Chr.16: 3582598-3582599 on Build GRCh38
rs926700658 Chr.16: 3582463-3582463 on Build GRCh38
rs755353749 Chr.16: 3582626-3582626 on Build GRCh38
rs201687045 Chr.16: 3582459-3582459 on Build GRCh38
rs768475866 Chr.16: 3582602-3582602 on Build GRCh38
rs531251083 Chr.16: 3582510-3582510 on Build GRCh38
rs775114324 Chr.16: 3582564-3582564 on Build GRCh38
rs556217576 Chr.16: 3582635-3582635 on Build GRCh38
rs773058133 Chr.16: 3582531-3582531 on Build GRCh38
rs745421287 Chr.16: 3582598-3582598 on Build GRCh38
rs377648201 Chr.16: 3582569-3582569 on Build GRCh38
rs748978984 Chr.16: 3582594-3582594 on Build GRCh38
rs769537789 Chr.16: 3582458-3582458 on Build GRCh38
rs778690592 Chr.16: 3582655-3582655 on Build GRCh38
rs771897046 Chr.16: 3582599-3582599 on Build GRCh38
rs527440287 Chr.16: 3582582-3582582 on Build GRCh38
rs368848191 Chr.16: 3582465-3582465 on Build GRCh38
rs200991968 Chr.16: 3582664-3582664 on Build GRCh38

View Full Product Details