The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001453.2 | NP_001444.2 | 1 - 1 | 1605 | 61 |
GenBank mRNA | BC134421.1 | - | 2 - 2 | 693 | 61 |