Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABBR1
Entrez Gene ID
2550
Gene Name
gamma-aminobutyric acid type B receptor subunit 1
Gene Aliases
GABABR1, GABBR1-3, GB1, GPRC3A
Chromosome Location
Chr.6: 29602228 - 29633135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319053.1 NP_001305982.1 9 - 10 1289 69
NM_001470.3 NP_001461.1 9 - 10 1353 69
NM_021903.2 NP_068703.1 4 - 5 1153 69
NM_021904.3 NP_068704.2 8 - 9 1167 69
XM_005248982.2 XP_005249039.1 9 - 10 1168 69
XM_006715047.3 XP_006715110.1 8 - 9 968 69
XM_011514453.2 XP_011512755.1 7 - 8 947 69
XM_011514455.1 XP_011512757.1 9 - 10 1168 69
XM_017010676.1 XP_016866165.1 5 - 6 699 69
GenBank mRNA AB208790.1 - 9 - 10 1088 69
AF099148.1 - 9 - 10 1175 69
AF301005.1 - 8 - 9 1070 69
AJ012185.1 - 8 - 9 1067 69
AJ012186.1 - 4 - 5 716 69
AJ012187.1 - 7 - 8 881 69
AJ012288.1 - 9 - 10 1110 69
AJ225028.1 - 9 - 10 1301 69
AJ225029.1 - 4 - 5 895 69
AK223619.1 - 4 - 5 938 69
AK290191.1 - 4 - 5 938 69
AK312308.1 - 4 - 5 934 69
BC041332.2 - 4 - 5 938 69
BC042598.1 - 4 - 5 927 69
BC050532.1 - 4 - 5 925 69
Y11044.1 - 8 - 9 1064 69

View Full Product Details