The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002208.4 | NP_002199.3 | 3 - 4 | 346 | 92 |
XM_011523823.2 | XP_011522125.1 | 3 - 4 | 784 | 92 | |
XM_011523825.2 | XP_011522127.1 | 3 - 4 | 782 | 92 | |
XM_011523827.2 | XP_011522129.1 | 3 - 4 | 776 | 92 | |
XM_011523828.2 | XP_011522130.1 | 3 - 4 | 780 | 92 | |
XM_017024586.1 | XP_016880075.1 | 4 - 5 | 2169 | 92 | |
XM_017024587.1 | XP_016880076.1 | 4 - 5 | 2169 | 92 | |
GenBank mRNA | BC113436.1 | - | 3 - 4 | 336 | 92 |
BC117207.1 | - | 3 - 4 | 336 | 92 | |
L25851.2 | - | 3 - 4 | 372 | 92 |