The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018911.2 | NP_061734.1 | 1 - 1 | 460 | 91 |
NM_031856.1 | NP_114062.1 | 1 - 1 | 460 | 91 | |
GenBank mRNA | AF152316.1 | - | 1 - 1 | 460 | 91 |
AF152486.1 | - | 1 - 1 | 460 | 91 | |
BC136749.1 | - | 1 - 1 | 555 | 91 | |
BC136750.1 | - | 1 - 1 | 555 | 91 |