Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196982930-196983173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR5
Entrez Gene ID
81494
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
UniGene
Hs.282594
Gene Chromosome Location
Chr.1: 196975022-197009725 on Build GRCh38
Transcripts
NM_030787.3 XM_011510020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627547 c.140C>G p.T47R Chr.1: 196982966-196982966 on Build GRCh38
159965 c.243G>A p.P81P Chr.1: 196983069-196983069 on Build GRCh38
227692 c.145G>A p.E49K Chr.1: 196982971-196982971 on Build GRCh38
3863973 c.256C>T p.P86S Chr.1: 196983010-196983010 on Build GRCh38
4527079 c.215G>A p.G72E Chr.1: 196982969-196982969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758679310 Chr.1: 196983095-196983095 on Build GRCh38
rs779001368 Chr.1: 196983020-196983020 on Build GRCh38
rs753989465 Chr.1: 196982980-196982980 on Build GRCh38
rs765238160 Chr.1: 196983091-196983091 on Build GRCh38
rs544857720 Chr.1: 196983068-196983068 on Build GRCh38
rs962677001 Chr.1: 196983014-196983014 on Build GRCh38
rs143503904 Chr.1: 196983039-196983039 on Build GRCh38
rs779854011 Chr.1: 196983107-196983107 on Build GRCh38
rs773840564 Chr.1: 196983045-196983045 on Build GRCh38
rs375843181 Chr.1: 196983032-196983032 on Build GRCh38
rs752717308 Chr.1: 196983092-196983092 on Build GRCh38
rs772279590 Chr.1: 196983086-196983086 on Build GRCh38
rs934983906 Chr.1: 196983078-196983078 on Build GRCh38
rs752894097 Chr.1: 196983021-196983021 on Build GRCh38
rs750169252 Chr.1: 196983045-196983047 on Build GRCh38
rs199913139 Chr.1: 196982971-196982971 on Build GRCh38
rs374676937 Chr.1: 196983096-196983096 on Build GRCh38
rs768077119 Chr.1: 196983043-196983043 on Build GRCh38
rs749182761 Chr.1: 196983112-196983112 on Build GRCh38
rs761567010 Chr.1: 196983052-196983052 on Build GRCh38
rs368326752 Chr.1: 196983041-196983041 on Build GRCh38
rs768577178 Chr.1: 196983122-196983122 on Build GRCh38
rs763857612 Chr.1: 196983031-196983031 on Build GRCh38
rs372885376 Chr.1: 196983079-196983079 on Build GRCh38
rs112201342 Chr.1: 196983063-196983063 on Build GRCh38
rs755844450 Chr.1: 196983102-196983102 on Build GRCh38
rs764958024 Chr.1: 196982968-196982969 on Build GRCh38
rs146025130 Chr.1: 196983058-196983058 on Build GRCh38
rs750756941 Chr.1: 196982972-196982972 on Build GRCh38
rs988214821 Chr.1: 196982981-196982981 on Build GRCh38
rs761721379 Chr.1: 196982966-196982968 on Build GRCh38
rs568332602 Chr.1: 196983139-196983139 on Build GRCh38
rs780377304 Chr.1: 196983126-196983126 on Build GRCh38
rs949171951 Chr.1: 196983093-196983093 on Build GRCh38
rs761319757 Chr.1: 196982974-196982974 on Build GRCh38
rs45489701 Chr.1: 196983138-196983138 on Build GRCh38
rs370641856 Chr.1: 196983004-196983004 on Build GRCh38
rs750261960 Chr.1: 196983097-196983097 on Build GRCh38
rs12097550 Chr.1: 196982962-196982962 on Build GRCh38
rs986414453 Chr.1: 196983038-196983038 on Build GRCh38
rs761513666 Chr.1: 196982964-196982964 on Build GRCh38
rs7532068 Chr.1: 196983069-196983069 on Build GRCh38
rs760047371 Chr.1: 196983090-196983090 on Build GRCh38
rs141256552 Chr.1: 196983059-196983059 on Build GRCh38

View Full Product Details