The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002795.3 | NP_002786.2 | 4 - 5 | 590 | 138 |
NR_104195.1 | 4 - 5 | 513 | 138 | ||
GenBank mRNA | BC013008.2 | - | 4 - 5 | 553 | 138 |
BQ428285.1 | - | 4 - 5 | 473 | 138 | |
D26598.1 | - | 4 - 5 | 493 | 138 |