The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015508.4 | NP_056323.2 | 1 - 2 | 381 | 63 |
GenBank mRNA | AK291261.1 | - | 1 - 2 | 216 | 63 |
BC050350.1 | - | 1 - 2 | 188 | 63 | |
BX537965.1 | - | 1 - 2 | 208 | 63 | |
DB033700.1 | - | 1 - 2 | 381 | 63 |