Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIPARP
Entrez Gene ID
25976
Gene Name
TCDD inducible poly(ADP-ribose) polymerase
Gene Aliases
ARTD14, PARP7, pART14
Chromosome Location
Chr.3: 156674416 - 156706770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184717.1 NP_001171646.1 2 - 3 1430 116
NM_001184718.1 NP_001171647.1 2 - 3 995 116
NM_015508.4 NP_056323.2 2 - 3 1345 116
GenBank mRNA AK097785.1 - 2 - 3 1174 116
AK291261.1 - 2 - 3 1180 116
BC034397.1 - 2 - 3 995 116
BC050350.1 - 2 - 3 1152 116
BX537965.1 - 2 - 3 1172 116
CR749647.1 - 2 - 3 1431 116

View Full Product Details