The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014434.1 | NP_001014434.1 | 4 - 5 | 729 | 63 |
NM_020204.2 | NP_064589.2 | 3 - 4 | 1166 | 63 | |
XM_005245350.3 | XP_005245407.2 | 4 - 5 | 1466 | 63 | |
XM_011509781.2 | XP_011508083.2 | 4 - 5 | 1472 | 63 | |
XM_017001849.1 | XP_016857338.1 | 4 - 5 | 1061 | 63 | |
GenBank mRNA | AJ277915.1 | - | 4 - 5 | 702 | 63 |
AK097614.1 | - | 2 - 3 | 933 | 63 | |
AY273889.2 | - | 3 - 4 | 1166 | 63 | |
BC131622.1 | - | 3 - 4 | 778 | 63 |