Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOP1
Entrez Gene ID
133060
Gene Name
otopetrin 1
Gene Aliases
-
Chromosome Location
Chr.4: 4188803 - 4226894 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177998.1 NP_819056.1 1 - 2 432 154
GenBank mRNA BC130430.1 - 1 - 2 427 154
BC130432.1 - 1 - 2 427 154

View Full Product Details