The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178844.3 | NP_849172.2 | 15 - 16 | 3189 | 65 |
NR_075083.2 | 16 - 17 | 3386 | 65 | ||
XM_017023027.1 | XP_016878516.1 | 15 - 16 | 3219 | 65 | |
XM_017023028.1 | XP_016878517.1 | 16 - 17 | 3393 | 65 | |
XM_017023029.1 | XP_016878518.1 | 17 - 18 | 3474 | 65 | |
XM_017023030.1 | XP_016878519.1 | 17 - 18 | 3535 | 65 | |
XM_017023031.1 | XP_016878520.1 | 15 - 16 | 3280 | 65 | |
XM_017023032.1 | XP_016878521.1 | 17 - 18 | 4236 | 65 | |
XM_017023033.1 | XP_016878522.1 | 17 - 18 | 3413 | 65 | |
XM_017023034.1 | XP_016878523.1 | 16 - 17 | 3332 | 65 | |
XM_017023035.1 | XP_016878524.1 | 16 - 17 | 3361 | 65 | |
XM_017023036.1 | XP_016878525.1 | 15 - 16 | 3920 | 65 | |
XM_017023037.1 | XP_016878526.1 | 16 - 17 | 3114 | 65 | |
XM_017023038.1 | XP_016878527.1 | 16 - 17 | 3087 | 65 | |
GenBank mRNA | AK074109.1 | - | 11 - 12 | 1067 | 65 |
AK090431.1 | - | 16 - 17 | 3382 | 65 | |
AY601811.1 | - | 15 - 16 | 3176 | 65 | |
FJ889357.1 | - | 12 - 13 | 2765 | 65 |