Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLRC3
Entrez Gene ID
197358
Gene Name
NLR family CARD domain containing 3
Gene Aliases
CLR16.2, NOD3
Chromosome Location
Chr.16: 3538272 - 3578373 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178844.3 NP_849172.2 4 - 5 600 105
NR_075083.2 4 - 5 661 105
XM_017023038.1 XP_016878527.1 4 - 5 603 105
GenBank mRNA AK090431.1 - 4 - 5 657 105
AK090476.1 - 4 - 5 600 105
AY601811.1 - 4 - 5 587 105
FJ889357.1 - 1 - 2 176 105

View Full Product Details