The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032477.2 | NP_115866.1 | 1 - 2 | 122 | 152 |
GenBank mRNA | AI091177.1 | - | 1 - 2 | 122 | 152 |
BC040035.1 | - | 1 - 2 | 97 | 152 | |
BM919526.1 | - | - | 102 | 152 |