The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001971.5 | NP_001962.3 | 3 - 4 | 240 | 106 |
GenBank mRNA | AF120493.1 | - | 3 - 4 | 240 | 106 |
BC069454.1 | - | 3 - 4 | 240 | 106 | |
BC075091.2 | - | 3 - 4 | 221 | 106 |