Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location
Chr.8: 52110832 - 52409879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014682.2 NP_055497.1 14 - 15 2415 93
XM_006716487.1 XP_006716550.1 14 - 15 2182 93
XM_011517629.1 XP_011515931.1 15 - 16 2338 93
XM_011517631.1 XP_011515933.1 16 - 17 2379 93
XM_011517632.1 XP_011515934.1 14 - 15 2193 93
XM_011517633.1 XP_011515935.1 14 - 15 2186 93
XM_011517634.1 XP_011515936.1 14 - 15 2382 93
XM_011517635.1 XP_011515937.1 14 - 15 2386 93
XM_011517636.2 XP_011515938.1 13 - 14 2463 93
XM_011517637.1 XP_011515939.1 12 - 13 1920 93
XM_011517638.2 XP_011515940.1 11 - 12 1961 93
XM_011517641.1 XP_011515943.1 16 - 17 2426 93
XM_011517642.1 XP_011515944.1 15 - 16 2338 93
XM_017014047.1 XP_016869536.1 14 - 15 2542 93
XM_017014048.1 XP_016869537.1 14 - 15 2546 93
XM_017014049.1 XP_016869538.1 15 - 16 2592 93
XM_017014050.1 XP_016869539.1 14 - 15 2379 93
XM_017014051.1 XP_016869540.1 13 - 14 2386 93
XM_017014052.1 XP_016869541.1 15 - 16 2596 93
XM_017014053.1 XP_016869542.1 15 - 16 2583 93
XM_017014054.1 XP_016869543.1 13 - 14 2632 93
XM_017014055.1 XP_016869544.1 13 - 14 2223 93
XM_017014056.1 XP_016869545.1 16 - 17 2633 93
XM_017014057.1 XP_016869546.1 14 - 15 2436 93
XM_017014058.1 XP_016869547.1 14 - 15 2197 93
XM_017014059.1 XP_016869548.1 14 - 15 2383 93
XM_017014060.1 XP_016869549.1 15 - 16 2420 93
XM_017014061.1 XP_016869550.1 13 - 14 2555 93
XM_017014062.1 XP_016869551.1 12 - 13 2307 93
XM_017014063.1 XP_016869552.1 13 - 14 2853 93
XM_017014064.1 XP_016869553.1 13 - 14 2467 93
XM_017014065.1 XP_016869554.1 15 - 16 2587 93
XM_017014066.1 XP_016869555.1 13 - 14 2226 93
XM_017014067.1 XP_016869556.1 13 - 14 2636 93
XM_017014068.1 XP_016869557.1 14 - 15 2427 93
XM_017014069.1 XP_016869558.1 11 - 12 2124 93
XM_017014070.1 XP_016869559.1 10 - 11 2045 93
XM_017014071.1 XP_016869560.1 11 - 12 2293 93
XM_017014072.1 XP_016869561.1 15 - 16 2457 93
XM_017014073.1 XP_016869562.1 15 - 16 2095 93
XM_017014074.1 XP_016869563.1 14 - 15 2378 93
XM_017014075.1 XP_016869564.1 15 - 16 2629 93
XM_017014076.1 XP_016869565.1 15 - 16 2633 93
XM_017014077.1 XP_016869566.1 15 - 16 2270 93
XM_017014078.1 XP_016869567.1 14 - 15 2550 93
XM_017014079.1 XP_016869568.1 14 - 15 2941 93
XM_017014080.1 XP_016869569.1 12 - 13 2182 93
XM_017014081.1 XP_016869570.1 14 - 15 2549 93
XM_017014082.1 XP_016869571.1 14 - 15 2384 93
XM_017014083.1 XP_016869572.1 13 - 14 2224 93
XM_017014084.1 XP_016869573.1 11 - 12 1962 93
GenBank mRNA AB011107.1 - 14 - 15 2279 93
BC031075.1 - 14 - 15 2312 93
BC117147.1 - 9 - 10 1672 93
BC117149.1 - 9 - 10 1672 93
BC143872.1 - 9 - 10 1667 93
BC143873.1 - 10 - 11 1760 93

View Full Product Details