Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location
Chr.8: 52110832 - 52409879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014682.2 NP_055497.1 23 - 24 3576 148
XM_006716487.1 XP_006716550.1 23 - 24 3343 148
XM_011517629.1 XP_011515931.1 24 - 25 3499 148
XM_011517631.1 XP_011515933.1 25 - 26 3540 148
XM_011517632.1 XP_011515934.1 23 - 24 3354 148
XM_011517633.1 XP_011515935.1 23 - 24 3347 148
XM_011517634.1 XP_011515936.1 23 - 24 3543 148
XM_011517635.1 XP_011515937.1 23 - 24 3547 148
XM_011517636.2 XP_011515938.1 22 - 23 3624 148
XM_011517637.1 XP_011515939.1 21 - 22 3081 148
XM_011517638.2 XP_011515940.1 20 - 21 3122 148
XM_011517641.1 XP_011515943.1 25 - 26 3587 148
XM_011517642.1 XP_011515944.1 23 - 24 3253 148
XM_017014047.1 XP_016869536.1 23 - 24 3703 148
XM_017014048.1 XP_016869537.1 23 - 24 3707 148
XM_017014049.1 XP_016869538.1 24 - 25 3753 148
XM_017014050.1 XP_016869539.1 23 - 24 3540 148
XM_017014051.1 XP_016869540.1 22 - 23 3547 148
XM_017014052.1 XP_016869541.1 24 - 25 3757 148
XM_017014053.1 XP_016869542.1 24 - 25 3744 148
XM_017014054.1 XP_016869543.1 22 - 23 3793 148
XM_017014055.1 XP_016869544.1 22 - 23 3384 148
XM_017014056.1 XP_016869545.1 25 - 26 3794 148
XM_017014057.1 XP_016869546.1 23 - 24 3597 148
XM_017014058.1 XP_016869547.1 23 - 24 3358 148
XM_017014059.1 XP_016869548.1 23 - 24 3544 148
XM_017014060.1 XP_016869549.1 24 - 25 3581 148
XM_017014061.1 XP_016869550.1 22 - 23 3716 148
XM_017014062.1 XP_016869551.1 21 - 22 3468 148
XM_017014063.1 XP_016869552.1 22 - 23 4014 148
XM_017014064.1 XP_016869553.1 22 - 23 3628 148
XM_017014065.1 XP_016869554.1 24 - 25 3748 148
XM_017014066.1 XP_016869555.1 22 - 23 3387 148
XM_017014067.1 XP_016869556.1 22 - 23 3797 148
XM_017014068.1 XP_016869557.1 23 - 24 3588 148
XM_017014069.1 XP_016869558.1 20 - 21 3285 148
XM_017014070.1 XP_016869559.1 19 - 20 3206 148
XM_017014071.1 XP_016869560.1 20 - 21 3454 148
XM_017014072.1 XP_016869561.1 24 - 25 3618 148
XM_017014073.1 XP_016869562.1 24 - 25 3256 148
XM_017014074.1 XP_016869563.1 23 - 24 3539 148
XM_017014075.1 XP_016869564.1 24 - 25 3790 148
XM_017014076.1 XP_016869565.1 24 - 25 3794 148
XM_017014077.1 XP_016869566.1 24 - 25 3431 148
XM_017014078.1 XP_016869567.1 23 - 24 3711 148
XM_017014079.1 XP_016869568.1 23 - 24 4102 148
XM_017014080.1 XP_016869569.1 21 - 22 3343 148
XM_017014081.1 XP_016869570.1 22 - 23 3464 148
XM_017014082.1 XP_016869571.1 22 - 23 3299 148
XM_017014083.1 XP_016869572.1 21 - 22 3139 148
XM_017014084.1 XP_016869573.1 19 - 20 2877 148
GenBank mRNA AB011107.1 - 23 - 24 3440 148
BC117147.1 - 18 - 19 2833 148
BC117149.1 - 18 - 19 2833 148
BC143872.1 - 18 - 19 2828 148
BC143873.1 - 19 - 20 2921 148

View Full Product Details