Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB11FIP3
Entrez Gene ID
9727
Gene Name
RAB11 family interacting protein 3
Gene Aliases
CART1, Rab11-FIP3
Chromosome Location
Chr.16: 425621 - 522485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142272.1 NP_001135744.1 5 - 6 687 55
NM_014700.3 NP_055515.1 7 - 8 1733 55
XM_005255713.3 XP_005255770.1 8 - 9 1903 55
XM_005255714.1 XP_005255771.1 6 - 7 748 55
XM_005255715.4 XP_005255772.1 6 - 7 1554 55
XM_011522764.2 XP_011521066.1 6 - 7 780 55
XM_011522765.2 XP_011521067.1 6 - 7 606 55
GenBank mRNA AB014565.1 - 7 - 8 1781 55
AF395731.1 - 7 - 8 1781 55
AK293644.1 - 5 - 6 602 55
AK303061.1 - 5 - 6 691 55
BC051360.1 - 7 - 8 1732 55

View Full Product Details