Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYS2
Entrez Gene ID
2998
Gene Name
glycogen synthase 2
Gene Aliases
-
Chromosome Location
Chr.12: 21536189 - 21604858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021957.3 NP_068776.2 7 - 8 1322 130
XM_006719063.3 XP_006719126.1 6 - 7 854 130
XM_017019245.1 XP_016874734.1 7 - 8 1333 130
GenBank mRNA AK308373.1 - 7 - 8 1294 130
BC126310.1 - 7 - 8 1092 130
BC126312.1 - 7 - 8 1092 130
S70004.1 - 7 - 8 1095 130

View Full Product Details