Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA5B1
Entrez Gene ID
127731
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
-
Chromosome Location
Chr.1: 20290907 - 20360442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039500.2 NP_001034589.2 17 - 18 2962 77
XM_006710360.3 XP_006710423.2 14 - 15 2237 77
XM_011540683.2 XP_011538985.1 17 - 18 2924 77
XM_011540684.2 XP_011538986.1 17 - 18 3049 77
XM_011540685.2 XP_011538987.1 17 - 18 3061 77
XM_011540688.2 XP_011538990.1 17 - 18 2924 77
XM_011540690.2 XP_011538992.1 16 - 17 2747 77
XM_011540691.2 XP_011538993.1 16 - 17 2738 77
XM_011540692.2 XP_011538994.1 17 - 18 2918 77
XM_011540693.1 XP_011538995.1 17 - 18 2924 77
XM_011540697.2 XP_011538999.1 11 - 12 2295 77
XM_017000305.1 XP_016855794.1 11 - 12 2041 77
XM_017000306.1 XP_016855795.1 12 - 13 2202 77
GenBank mRNA AK057346.1 - 6 - 7 1045 77
AK295797.1 - 17 - 18 2946 77
BC101378.1 - 6 - 7 1042 77
BC101379.2 - 6 - 7 1042 77
BC101380.1 - 6 - 7 1042 77
BC101381.1 - 6 - 7 1042 77

View Full Product Details